Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- eif4a1a
- Entrez Gene ID
-
386634
- Gene Name
- eukaryotic translation initiation factor 4A1A
- Gene Aliases
-
eif4a1, fb49a04, im:7143023, wu:fb20a10, wu:fb49a04, wu:fc76a02, wu:fc96c01, wu:fd15g03
- Chromosome Location
-
Chr.7: 26510532 - 26518089 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198366.2 |
NP_938180.1 |
8 - 9 |
942 |
91 |
GenBank mRNA |
BC048899.1 |
- |
8 - 9 |
911 |
91 |
View Full Product Details