Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ik
Entrez Gene ID
322241
Gene Name
IK cytokine
Gene Aliases
wu:fb54c03, zgc:56500
Chromosome Location
Chr.21: 44013487 - 44037107 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199589.1 NP_955883.1 17 - 18 1545 73
GenBank mRNA BC049322.1 - 17 - 18 1545 73

View Full Product Details