Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ccdc28a
Entrez Gene ID
326808
Gene Name
coiled-coil domain containing 28A
Gene Aliases
wu:fe24b05, zgc:63962
Chromosome Location
Chr.17: 33409032 - 33416091 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199748.1 NP_956042.1 2 - 3 266 88
XM_005158720.4 XP_005158777.1 2 - 3 475 88
GenBank mRNA BC053169.1 - 2 - 3 266 88

View Full Product Details