Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
utp11
Entrez Gene ID
336149
Gene Name
UTP11 small subunit processome component
Gene Aliases
CGI-94, fj52a01, utp11l, wu:fj52a01, zgc:56621
Chromosome Location
Chr.16: 404336 - 409231 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199998.1 NP_956292.1 6 - 7 687 110
GenBank mRNA BC049053.1 - 6 - 7 687 110

View Full Product Details