Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lum
Entrez Gene ID
415149
Gene Name
lumican
Gene Aliases
zgc:86661
Chromosome Location
Chr.4: 16351388 - 16354268 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002059.1 NP_001002059.1 2 - 3 937 80
GenBank mRNA BC071347.1 - 2 - 3 937 80
CU468778.1 - 1 - 2 879 80

View Full Product Details