Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snrnp27
Entrez Gene ID
436976
Gene Name
small nuclear ribonucleoprotein 27 (U4/U6.U5)
Gene Aliases
zgc:92615
Chromosome Location
Chr.5: 13767743 - 13778726 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002703.2 NP_001002703.1 2 - 3 268 61
GenBank mRNA BC076105.1 - 2 - 3 269 61

View Full Product Details