Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snrnp27
Entrez Gene ID
436976
Gene Name
small nuclear ribonucleoprotein 27 (U4/U6.U5)
Gene Aliases
zgc:92615
Chromosome Location
Chr.5: 13767743 - 13778726 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002703.2 NP_001002703.1 4 - 5 467 120
GenBank mRNA BC076105.1 - 4 - 5 468 120

View Full Product Details