Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lrrc23
Entrez Gene ID
450033
Gene Name
leucine rich repeat containing 23
Gene Aliases
zgc:101782
Chromosome Location
Chr.23: 20127883 - 20133982 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006054.1 NP_001006054.1 6 - 7 1221 146
GenBank mRNA BC083278.1 - 6 - 7 1221 146

View Full Product Details