Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc50a1
Entrez Gene ID
503533
Gene Name
solute carrier family 50 member 1
Gene Aliases
rag1ap1, zgc:113092
Chromosome Location
Chr.16: 23303894 - 23315610 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012497.1 NP_001012515.1 2 - 3 298 134
GenBank mRNA BC090168.1 - 2 - 3 298 134

View Full Product Details