Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
afg1lb
Entrez Gene ID
555926
Gene Name
AFG1 like ATPase b
Gene Aliases
lace1, lace1b, si:ch211-278n15.1
Chromosome Location
Chr.20: 32337364 - 32405428 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025482.2 NP_001020653.2 5 - 6 724 90
XM_021468423.1 XP_021324098.1 4 - 5 90
GenBank mRNA BC115066.1 - 5 - 6 724 90

View Full Product Details