Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gigyf2
Entrez Gene ID
561830
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
fb57e02, gigfy2, im:6911549, wu:fb57e02, zgc:111944
Chromosome Location
Chr.2: 279401 - 301823 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030161.2 NP_001025332.2 20 - 21 2491 73
XM_017358654.2 XP_017214143.1 20 - 21 2582 73
GenBank mRNA BC098603.1 - 20 - 21 2509 73

View Full Product Details