Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ptpn23b
Entrez Gene ID
560417
Gene Name
protein tyrosine phosphatase, non-receptor type 23, b
Gene Aliases
ptp23b, si:dkeyp-114f9.2
Chromosome Location
Chr.19: 19241341 - 19271270 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044928.1 NP_001038393.1 10 - 11 1079 151

View Full Product Details