Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrpl37
Entrez Gene ID
567819
Gene Name
mitochondrial ribosomal protein L37
Gene Aliases
im:6895748, zgc:162947
Chromosome Location
Chr.2: 10914039 - 10931770 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082855.1 NP_001076324.1 1 - 2 494 127
GenBank mRNA BC134163.1 - 1 - 2 494 127

View Full Product Details