Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mitfa
Entrez Gene ID
30080
Gene Name
melanocyte inducing transcription factor a
Gene Aliases
nacre, z3A.1
Chromosome Location
Chr.6: 43399763 - 43433295 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001362262.1 NP_001349191.1 7 - 8 840 78
NM_130923.2 NP_570998.1 4 - 5 546 78
XM_021476774.1 XP_021332449.1 4 - 5 611 78
XM_021476775.1 XP_021332450.1 5 - 6 721 78
GenBank mRNA AF119220.1 - 4 - 5 546 78
BC056318.1 - 4 - 5 538 78
BC065647.1 - 4 - 5 572 78

View Full Product Details