Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- inab
- Entrez Gene ID
-
30230
- Gene Name
- internexin neuronal intermediate filament protein, alpha b
- Gene Aliases
-
gefiltin, zgc:110516
- Chromosome Location
-
Chr.1: 31864468 - 31867717 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U89710.1 |
- |
1 - 2 |
1044 |
60 |
View Full Product Details