Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rx2
Entrez Gene ID
30473
Gene Name
retinal homeobox gene 2
Gene Aliases
-
Chromosome Location
Chr.2: 55833425 - 55861351 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131226.2 NP_571301.2 2 - 3 641 136
GenBank mRNA AF001908.1 - 1 - 2 290 136
BC095004.1 - 2 - 3 641 136
EH590819.1 - 2 - 3 482 136

View Full Product Details