Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stat1a
Entrez Gene ID
30768
Gene Name
signal transducer and activator of transcription 1a
Gene Aliases
stat1
Chromosome Location
Chr.22: 12798626 - 12825110 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131480.1 NP_571555.1 18 - 19 1731 66
GenBank mRNA AJ005692.1 - 18 - 19 1731 66
BC045863.1 - 17 - 18 1878 66
BC060933.1 - 18 - 19 1760 66

View Full Product Details