Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
csf1ra
Entrez Gene ID
64274
Gene Name
colony stimulating factor 1 receptor, a
Gene Aliases
CSF-1R, M-CSF-R, csf1r, fms, panther
Chromosome Location
Chr.14: 3143344 - 3182333 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131672.1 NP_571747.1 5 - 6 1025 64
XM_021481056.1 XP_021336731.1 6 - 7 64
XM_021481057.1 XP_021336732.1 6 - 7 64
GenBank mRNA AF240639.1 - 5 - 6 1025 64
BC162803.1 - 5 - 6 960 64
CU458899.1 - 5 - 6 64

View Full Product Details