Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tpbga
Entrez Gene ID
373095
Gene Name
trophoblast glycoprotein a
Gene Aliases
YF-296, sb:cb613, tpbgl, waif1a, zgc:109764
Chromosome Location
Chr.19: 14059349 - 14061987 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194392.2 NP_919373.2 1 - 2 435 74
GenBank mRNA AB097825.1 - 1 - 2 435 74
BC099984.1 - 1 - 2 323 74
CU468751.1 - 1 - 2 267 74
JN653073.1 - 1 - 2 435 74

View Full Product Details