Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mboat7
Entrez Gene ID
393509
Gene Name
membrane bound O-acyltransferase domain containing 7
Gene Aliases
leng4, zgc:66032
Chromosome Location
Chr.16: 32014555 - 32026450 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200537.1 NP_956831.1 7 - 8 919 87
XM_021466647.1 XP_021322322.1 7 - 8 1014 87
GenBank mRNA BC056305.1 - 7 - 8 919 87

View Full Product Details