Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col1a1b
Entrez Gene ID
325675
Gene Name
collagen, type I, alpha 1b
Gene Aliases
alpha3(I), col1a3, fj59a10, hm:zeh0348, wu:fa95h05, wu:fd02a10, wu:fj59a10
Chromosome Location
Chr.12: 3095617 - 3133500 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201478.1 NP_958886.1 44 - 45 3308 84
XM_005155976.4 XP_005156033.1 44 - 45 3351 84
XM_005155980.4 XP_005156037.1 42 - 43 3192 84
GenBank mRNA AJ318214.1 - 3 - 4 185 84
BC058045.1 - 44 - 45 3308 84
BC066384.1 - 44 - 45 84

View Full Product Details