Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lpgat1
Entrez Gene ID
561832
Gene Name
lysophosphatidylglycerol acyltransferase 1
Gene Aliases
fc21c12, fj66b10, si:dkey-30h14.2, wu:fc21c12, wu:fj66b10, zgc:112278
Chromosome Location
Chr.20: 13781600 - 13874344 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029958.1 NP_001025129.1 7 - 8 1024 86
XM_005160439.4 XP_005160496.1 7 - 8 1436 86
GenBank mRNA BC098616.1 - 7 - 8 1024 86

View Full Product Details