Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
smyd1b
Entrez Gene ID
569027
Gene Name
SET and MYND domain containing 1b
Gene Aliases
sth, symd1
Chromosome Location
Chr.8: 1015243 - 1051544 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039636.1 NP_001034725.1 6 - 7 882 96
XM_021478292.1 XP_021333967.1 5 - 6 949 96
GenBank mRNA BC115351.1 - 6 - 7 886 96
BC163095.1 - 5 - 6 932 96
DQ323979.1 - 6 - 7 882 96
DQ323980.1 - 5 - 6 843 96
DQ851819.1 - 2 - 3 256 96

View Full Product Details