Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cnpy3
Entrez Gene ID
550136
Gene Name
canopy FGF signaling regulator 3
Gene Aliases
im:6892510, tnrc5
Chromosome Location
Chr.1: 10462692 - 10473588 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039805.2 NP_001034894.2 2 - 3 299 87
GenBank mRNA AB201385.1 - 2 - 3 327 87
BC083297.1 - 2 - 3 341 87
BC133981.1 - 2 - 3 299 87

View Full Product Details