Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
illr3
Entrez Gene ID
677740
Gene Name
immune-related, lectin-like receptor 3
Gene Aliases
-
Chromosome Location
Chr.19: 3266957 - 3272998 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040039.1 NP_001035128.1 5 - 6 454 100
NM_001044742.1 NP_001038207.1 4 - 5 355 100
XM_021468134.1 XP_021323809.1 5 - 6 481 100
GenBank mRNA AY986756.1 - 5 - 6 454 100
AY986757.1 - 4 - 5 355 100
BC163143.1 - 4 - 5 355 100
BC163144.1 - 4 - 5 355 100

View Full Product Details