Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab40c
Entrez Gene ID
678600
Gene Name
RAB40c, member RAS oncogene family
Gene Aliases
cb453, wu:fk50d06, zgc:136966
Chromosome Location
Chr.3: 26751861 - 26787196 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040348.1 NP_001035438.1 5 - 6 662 67
GenBank mRNA BC115347.1 - 5 - 6 662 67

View Full Product Details