Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gas2l3
Entrez Gene ID
566151
Gene Name
growth arrest-specific 2 like 3
Gene Aliases
fc11d07, si:ch211-203h15.1, wu:fc11d07
Chromosome Location
Chr.4: 1686621 - 1720648 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045098.1 NP_001038563.1 4 - 5 487 81
XM_005164456.4 XP_005164513.2 4 - 5 544 81
XM_021475058.1 XP_021330733.1 4 - 5 455 81

View Full Product Details