Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spata5l1
Entrez Gene ID
767648
Gene Name
spermatogenesis associated 5-like 1
Gene Aliases
wu:fi26h05, wu:fi39b02, zgc:153294
Chromosome Location
Chr.18: 5530083 - 5542367 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076588.2 NP_001070056.2 5 - 6 1464 93
GenBank mRNA BC124305.1 - 5 - 6 1485 93

View Full Product Details