Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fdx1b
Entrez Gene ID
562135
Gene Name
ferredoxin 1b
Gene Aliases
si:rp71-5c20.1
Chromosome Location
Chr.5: 171256 - 210554 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100949.1 NP_001094419.1 3 - 4 534 111

View Full Product Details