Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cry4
Entrez Gene ID
83775
Gene Name
cryptochrome circadian regulator 4
Gene Aliases
fi23e01, wu:fi23e01, zcry4
Chromosome Location
Chr.22: 819650 - 834105 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131787.1 NP_571862.1 9 - 10 1485 53
XM_005168277.4 XP_005168334.1 10 - 11 1712 53
XM_009295766.3 XP_009294041.1 10 - 11 1699 53
GenBank mRNA AB042253.1 - 9 - 10 1485 53
BC097079.1 - 8 - 9 1229 53

View Full Product Details