The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144764.2 | NP_658910.2 | 15 - 16 | 1498 | 110 |
GenBank mRNA | AJ306396.1 | - | 15 - 16 | 1501 | 110 |
BC121194.1 | - | 15 - 16 | 1442 | 110 |