Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rbpja
Entrez Gene ID
386772
Gene Name
recombination signal binding protein for immunoglobulin kappa J region a
Gene Aliases
Su(H), Su(H)1, Su(h)A, ik:tdsubc_1e8, rbpj, rbpsuh, tdsubc_1e8, xx:tdsubc_1e8
Chromosome Location
Chr.1: 14454663 - 14464020 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198878.2 NP_942579.2 8 - 9 916 147
GenBank mRNA AY128532.1 - 8 - 9 919 150
AY332617.1 - 8 - 9 889 147
BC163922.1 - 8 - 9 916 147

View Full Product Details