Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
chchd10
Entrez Gene ID
393757
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 10
Gene Aliases
chchd2, zgc:73308
Chromosome Location
Chr.21: 15704655 - 15708626 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200784.1 NP_957078.1 2 - 3 351 67
GenBank mRNA BC059631.1 - 2 - 3 351 67

View Full Product Details