Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arl2bp
Entrez Gene ID
393976
Gene Name
ADP-ribosylation factor-like 2 binding protein
Gene Aliases
BART1, zgc:66308, zgc:77496
Chromosome Location
Chr.18: 167459 - 171697 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201001.2 NP_957295.2 2 - 3 154 81
GenBank mRNA AB128034.1 - 1 - 2 44 81
BC054678.1 - 2 - 3 167 81
BC066483.1 - 2 - 3 106 81
EH541682.1 - 2 - 3 177 81

View Full Product Details