Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ndel1a
Entrez Gene ID
394244
Gene Name
nudE neurodevelopment protein 1-like 1a
Gene Aliases
-
Chromosome Location
Chr.12: 26467821 - 26487363 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201344.1 NP_958501.1 7 - 8 902 81
XM_009306604.3 XP_009304879.1 7 - 8 928 81
XM_009306605.2 XP_009304880.1 7 - 8 928 81
GenBank mRNA BC055587.1 - 7 - 8 902 81
BC164385.1 - 6 - 7 829 81
EF133514.1 - 7 - 8 830 81

View Full Product Details