Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
uap1
Entrez Gene ID
322513
Gene Name
UDP-N-acetylglucosamine pyrophosphorylase 1
Gene Aliases
fb63d12, wu:fb63d12, zgc:55280
Chromosome Location
Chr.6: 35036189 - 35052282 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212621.2 NP_997786.2 8 - 9 1517 85
GenBank mRNA BC044137.1 - 8 - 9 1463 85
BC164218.1 - 7 - 8 1395 85

View Full Product Details