Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trim55a
Entrez Gene ID
436631
Gene Name
tripartite motif containing 55a
Gene Aliases
trim55, wu:fb71c01, zgc:92123
Chromosome Location
Chr.2: 42114310 - 42128660 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002358.1 NP_001002358.1 5 - 6 887 109
GenBank mRNA BC075897.1 - 5 - 6 887 109

View Full Product Details