Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc25a36a
Entrez Gene ID
436940
Gene Name
solute carrier family 25 member 36a
Gene Aliases
zgc:92447
Chromosome Location
Chr.2: 264249 - 292024 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002667.1 NP_001002667.1 6 - 7 1106 62
XM_009298709.3 XP_009296984.1 3 - 4 3421 62
XM_009298710.3 XP_009296985.1 4 - 5 3304 62
XM_021479882.1 XP_021335557.1 7 - 8 1230 62
XM_021479889.1 XP_021335564.1 4 - 5 3282 62
GenBank mRNA BC076521.1 - 6 - 7 1106 62
BC164094.1 - 6 - 7 778 62

View Full Product Details