Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cd2ap
Entrez Gene ID
494040
Gene Name
CD2-associated protein
Gene Aliases
cd2apl, fj51c01, im:7153970, si:ch211-207n9.1, wu:fj51c01, zgc:103705
Chromosome Location
Chr.20: 35857355 - 35980223 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008583.2 NP_001008583.2 4 - 5 719 134
XM_005160684.4 XP_005160741.1 3 - 4 350 134
XM_009294709.3 XP_009292984.1 4 - 5 750 134
GenBank mRNA BC086846.1 - 3 - 4 578 134
BC164631.1 - 3 - 4 389 134

View Full Product Details