Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sinhcaf
Entrez Gene ID
563991
Gene Name
SIN3-HDAC complex associated factor
Gene Aliases
fam60a, si:ch211-11c20.3
Chromosome Location
Chr.4: 16635446 - 16644708 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025531.1 NP_001020702.1 3 - 4 259 101
GenBank mRNA BC162866.1 - 3 - 4 259 101
BC162884.1 - 3 - 4 259 101

View Full Product Details