Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ankha
Entrez Gene ID
571148
Gene Name
ANKH inorganic pyrophosphate transport regulator a
Gene Aliases
wu:fj64g09, zgc:110290
Chromosome Location
Chr.24: 10349147 - 10394291 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030259.2 NP_001025430.2 7 - 8 1219 72
XM_021470216.1 XP_021325891.1 5 - 6 897 72
GenBank mRNA BC096801.1 - 8 - 8 1526 72

View Full Product Details