Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pcxa
Entrez Gene ID
553283
Gene Name
pyruvate carboxylase a
Gene Aliases
PYCa, pcl, zgc:123047
Chromosome Location
Chr.21: 26733345 - 26966059 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328356.1 NP_001315285.1 4 - 5 798 56
GenBank mRNA BC086839.1 - 4 - 5 765 56
BC109463.1 - 4 - 5 809 56

View Full Product Details