Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atxn7l2a
Entrez Gene ID
692316
Gene Name
ataxin 7-like 2a
Gene Aliases
fi32b11, wu:fi32b11, zgc:136546
Chromosome Location
Chr.11: 331724 - 360263 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045285.1 NP_001038750.1 8 - 9 1349 98
XM_005172905.4 XP_005172962.1 8 - 9 1474 98
XM_005172906.3 XP_005172963.1 9 - 10 1642 98
GenBank mRNA BC056808.1 - 8 - 9 1368 98
BC116583.1 - 8 - 9 1349 98

View Full Product Details