Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prkar2aa
Entrez Gene ID
556042
Gene Name
protein kinase, cAMP-dependent, regulatory, type II, alpha A
Gene Aliases
wu:fb55f12, wu:fj29e12, wu:fj33e06, zgc:153742
Chromosome Location
Chr.8: 26330277 - 26369116 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077370.2 NP_001070838.2 5 - 6 1079 136
GenBank mRNA BC124725.1 - 5 - 6 1080 136
BC164493.1 - 5 - 6 719 136
BQ481277.1 - 2 - 3 195 136

View Full Product Details