Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
usp9
Entrez Gene ID
568683
Gene Name
ubiquitin specific peptidase 9
Gene Aliases
-
Chromosome Location
Chr.9: 33261200 - 33321037 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077449.1 NP_001070917.1 6 - 7 774 81
XM_005167736.4 XP_005167793.1 7 - 8 1256 81
XM_009304755.3 XP_009303030.1 7 - 8 1257 81
XM_009304756.3 XP_009303031.1 7 - 8 1257 81
XM_009304757.3 XP_009303032.1 7 - 8 1258 81
GenBank mRNA DQ086492.1 - 6 - 7 774 81

View Full Product Details