The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080174.1 | NP_001073643.1 | 6 - 7 | 926 | 64 |
XM_009303487.3 | XP_009301762.1 | 6 - 7 | 961 | 64 | |
XM_009303488.3 | XP_009301763.1 | 6 - 7 | 995 | 64 | |
GenBank mRNA | BC129442.1 | - | 6 - 7 | 926 | 64 |