Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kdm6al
Entrez Gene ID
569207
Gene Name
lysine (K)-specific demethylase 6A, like
Gene Aliases
utxl1, uty, zgc:158313
Chromosome Location
Chr.6: 59851523 - 59895080 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080193.1 NP_001073662.1 24 - 25 3521 72
XM_005168796.4 XP_005168853.1 25 - 26 6357 72
GenBank mRNA BC066564.1 - 24 - 25 4403 72
BC129198.1 - 24 - 25 3521 72

View Full Product Details