Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
antxr2b
Entrez Gene ID
560841
Gene Name
ANTXR cell adhesion molecule 2b
Gene Aliases
cmg2b
Chromosome Location
Chr.10: 252175 - 266250 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102568.1 NP_001096038.1 13 - 14 1076 94
GenBank mRNA EF591979.1 - 13 - 14 1076 94

View Full Product Details