Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kita
Entrez Gene ID
30256
Gene Name
KIT proto-oncogene, receptor tyrosine kinase a
Gene Aliases
c-kit, kit, receptor, sparse
Chromosome Location
Chr.20: 22331146 - 22378461 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131053.1 NP_571128.1 4 - 5 801 134
XM_009294634.3 XP_009292909.1 4 - 5 846 134
XM_009294635.3 XP_009292910.1 4 - 5 876 134
GenBank mRNA AF153446.1 - 4 - 5 801 134
BC162502.1 - 4 - 5 816 134
CU458898.1 - 4 - 5 752 134

View Full Product Details