Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sebox
Entrez Gene ID
282666
Gene Name
SEBOX homeobox
Gene Aliases
og9x
Chromosome Location
Chr.5: 67371650 - 67382183 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320052.1 NP_001306981.1 2 - 3 386 57
GenBank mRNA AF466189.1 - 2 - 3 386 57

View Full Product Details